Order Entry
Netherlands
ContactUsLinkComponent
Human TIM 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TIM 1 in tissue homogenates, urine, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3006-96
Supplier:  ANTIBODIES.COM
New Product
Human TIM 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TIM 1 in tissue homogenates, urine, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3006-96
Supplier:  ANTIBODIES.COM