Human Hepatitis B Virus Surface Antigen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Hepatitis B Virus Surface Antigen in serum, plasma, cell culture supernatant, ce 1 * 96 Tests
Catalog # ANTIA326612-96
Supplier: ANTIBODIES.COM