Order Entry
Netherlands
ContactUsLinkComponent
Human Stanniocalcin 2/STC-2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Stanniocalcin 2/STC-2 in serum, plasma, tissue homogenates, cell lysates, cell culture super 1 * 48 Tests
Catalog # ANTIA5746-48
Supplier:  Antibodies.com
New Product
Human Stanniocalcin 2/STC-2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Stanniocalcin 2/STC-2 in serum, plasma, tissue homogenates, cell lysates, cell culture super 1 * 48 Tests
Catalog # ANTIA5746-48
Supplier:  Antibodies.com