Order Entry
Netherlands
ContactUsLinkComponent
Rabbit IL-18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit IL-18 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4236-48
Supplier:  Antibodies.com
New Product
Rabbit IL-18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit IL-18 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4236-48
Supplier:  Antibodies.com