Order Entry
Netherlands
ContactUsLinkComponent
Rat Angiopoietin 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rat Angiopoietin 1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3374-48
Supplier:  Antibodies.com
New Product
Rat Angiopoietin 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rat Angiopoietin 1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3374-48
Supplier:  Antibodies.com