Order Entry
Netherlands
ContactUsLinkComponent
Human AIRE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human AIRE in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5054-48
Supplier:  Antibodies.com
New Product
Human AIRE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human AIRE in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5054-48
Supplier:  Antibodies.com