Order Entry
Netherlands
ContactUsLinkComponent
Mouse Albumin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse Albumin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4028-48
Supplier:  Antibodies.com
New Product
Mouse Albumin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse Albumin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4028-48
Supplier:  Antibodies.com