Order Entry
Netherlands
ContactUsLinkComponent
Rat SRD5A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat SRD5A1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5225-48
Supplier:  Antibodies.com
New Product
Rat SRD5A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat SRD5A1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5225-48
Supplier:  Antibodies.com