Order Entry
Netherlands
ContactUsLinkComponent
Human Inhibin beta A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Inhibin beta A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
  ANTIA3628-48
New Product
Human Inhibin beta A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Inhibin beta A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
  ANTIA3628-48