Order Entry
Netherlands
ContactUsLinkComponent
Mouse Estriol ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of m ouse Estriol in serum and plasma. 1 * 96 Tests
Catalog # ANTIA327142-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Estriol ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of m ouse Estriol in serum and plasma. 1 * 96 Tests
Catalog # ANTIA327142-96
Supplier:  ANTIBODIES.COM