Order Entry
Netherlands
ContactUsLinkComponent
Porcine IL-17A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of porcine IL-17A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA3582-48
Supplier:  Antibodies.com
New Product
Porcine IL-17A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of porcine IL-17A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA3582-48
Supplier:  Antibodies.com