Order Entry
Netherlands
ContactUsLinkComponent
Mouse GLP-1R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse GLP-1R in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5159-48
New Product
Mouse GLP-1R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse GLP-1R in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5159-48