Order Entry
Netherlands
ContactUsLinkComponent
Rat Glucokinase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Glucokinase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3863-48
Supplier:  Antibodies.com
New Product
Rat Glucokinase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Glucokinase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3863-48
Supplier:  Antibodies.com