Human Tankyrase 2/TNKS2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Tankyrase 2/TNKS2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 48 Tests
Catalog # ANTIA4862-48
Supplier: Antibodies.com