Order Entry
Netherlands
ContactUsLinkComponent
Human Biglycan ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Biglycan in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5494-48
Supplier:  Antibodies.com
New Product
Human Biglycan ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Biglycan in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5494-48
Supplier:  Antibodies.com