Order Entry
Netherlands
ContactUsLinkComponent
Human VRK1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human VRK1 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5191-48
Supplier:  Antibodies.com
New Product
Human VRK1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human VRK1 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5191-48
Supplier:  Antibodies.com