Order Entry
Netherlands
ContactUsLinkComponent
Zebrafish IGFBP1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of zebrafish IGFBP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA4072-96
Supplier:  ANTIBODIES.COM
New Product
Zebrafish IGFBP1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of zebrafish IGFBP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA4072-96
Supplier:  ANTIBODIES.COM