Order Entry
Netherlands
ContactUsLinkComponent
Human Mesothelin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Mesothelin in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA4895-48
New Product
Human Mesothelin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Mesothelin in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA4895-48