Order Entry
Netherlands
ContactUsLinkComponent
Human TIL/TLR1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human TIL/TLR1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3782-48
Supplier:  Antibodies.com
New Product
Human TIL/TLR1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human TIL/TLR1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3782-48
Supplier:  Antibodies.com