Order Entry
Netherlands
ContactUsLinkComponent
Mouse Synaptopodin 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Synaptopodin 2 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5499-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Synaptopodin 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Synaptopodin 2 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5499-96
Supplier:  ANTIBODIES.COM