Order Entry
Netherlands
ContactUsLinkComponent
Rat IGFBP6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat IGFBP6 in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA4204-48
New Product
Rat IGFBP6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat IGFBP6 in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA4204-48