Order Entry
Netherlands
ContactUsLinkComponent
Human mtTFA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human mtTFA in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5533-48
Supplier:  Antibodies.com
New Product
Human mtTFA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human mtTFA in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5533-48
Supplier:  Antibodies.com