Order Entry
Netherlands
ContactUsLinkComponent
Human Cytochrome P450 Reductase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Cytochrome P450 Reductase in tissue homogenates, cell lysates or other biological fluids 1 * 48 Tests
  ANTIA5616-48
New Product
Human Cytochrome P450 Reductase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Cytochrome P450 Reductase in tissue homogenates, cell lysates or other biological fluids 1 * 48 Tests
  ANTIA5616-48