Order Entry
Netherlands
ContactUsLinkComponent
Human Abi-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Abi-1 in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA6783-48
New Product
Human Abi-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Abi-1 in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA6783-48