Order Entry
Netherlands
ContactUsLinkComponent
Mouse Trehalase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Trehalase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6460-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Trehalase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Trehalase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6460-96
Supplier:  ANTIBODIES.COM