Order Entry
Netherlands
ContactUsLinkComponent
Human CYP26A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human CYP26A1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6146-48
Supplier:  ANTIBODIES.COM
New Product
Human CYP26A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human CYP26A1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6146-48
Supplier:  ANTIBODIES.COM