Order Entry
Netherlands
ContactUsLinkComponent
Human 4-1BBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human 4-1BBL in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7263-48
Supplier:  Antibodies.com
New Product
Human 4-1BBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human 4-1BBL in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7263-48
Supplier:  Antibodies.com