Order Entry
Netherlands
ContactUsLinkComponent
Human Fibroblast Activation Protein alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Fibroblast Activation Protein alpha in serum, plasma, tissue homogenates, cell 1 * 48 Tests
  ANTIA6527-48
New Product
Human Fibroblast Activation Protein alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Fibroblast Activation Protein alpha in serum, plasma, tissue homogenates, cell 1 * 48 Tests
  ANTIA6527-48