Order Entry
Netherlands
ContactUsLinkComponent
Human ASTL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human ASTL in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6519-48
Supplier:  Antibodies.com
New Product
Human ASTL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human ASTL in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6519-48
Supplier:  Antibodies.com