Order Entry
Netherlands
ContactUsLinkComponent
Human ENGASE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ENGASE in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7153-48
Supplier:  Antibodies.com
New Product
Human ENGASE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ENGASE in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7153-48
Supplier:  Antibodies.com