Order Entry
Netherlands
ContactUsLinkComponent
Mouse Peroxiredoxin 1/PAG ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Peroxiredoxin 1/PAG in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7181-48
Supplier:  Antibodies.com
New Product
Mouse Peroxiredoxin 1/PAG ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Peroxiredoxin 1/PAG in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7181-48
Supplier:  Antibodies.com