Order Entry
Netherlands
ContactUsLinkComponent
Human IREB2/IRP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human IREB2/IRP2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA7997-48
New Product
Human IREB2/IRP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human IREB2/IRP2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA7997-48