Order Entry
Netherlands
ContactUsLinkComponent
Porcine VEGF Receptor 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of porcine VEGF Receptor 2 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7934-48
Supplier:  Antibodies.com
New Product
Porcine VEGF Receptor 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of porcine VEGF Receptor 2 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7934-48
Supplier:  Antibodies.com