Order Entry
Netherlands
ContactUsLinkComponent
Human Sodium/Hydrogen Exchanger 1/NHE-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Sodium/Hydrogen Exchanger 1/NHE-1 in tissue homogenates, cell lysates or other b 1 * 48 Tests
Catalog # ANTIA6326-48
Supplier:  Antibodies.com
New Product
Human Sodium/Hydrogen Exchanger 1/NHE-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Sodium/Hydrogen Exchanger 1/NHE-1 in tissue homogenates, cell lysates or other b 1 * 48 Tests
Catalog # ANTIA6326-48
Supplier:  Antibodies.com