Order Entry
Netherlands
ContactUsLinkComponent
Human Cyclin B2/CCNB2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Cyclin B2/CCNB2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6669-48
Supplier:  Antibodies.com
New Product
Human Cyclin B2/CCNB2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Cyclin B2/CCNB2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6669-48
Supplier:  Antibodies.com