Order Entry
Netherlands
ContactUsLinkComponent
Human IL-12RB2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human IL-12RB2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6294-48
Supplier:  Antibodies.com
New Product
Human IL-12RB2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human IL-12RB2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6294-48
Supplier:  Antibodies.com