Order Entry
Netherlands
ContactUsLinkComponent
Human Noradrenaline Transporter ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Noradrenaline Transporter in tissue homogenates, cell lysates or other biological fluids 1 * 96 Tests
Catalog # ANTIA8018-96
Supplier:  ANTIBODIES.COM
New Product
Human Noradrenaline Transporter ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Noradrenaline Transporter in tissue homogenates, cell lysates or other biological fluids 1 * 96 Tests
Catalog # ANTIA8018-96
Supplier:  ANTIBODIES.COM