Order Entry
Netherlands
ContactUsLinkComponent
Human Rac1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human Rac1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7552-96
Supplier:  ANTIBODIES.COM
New Product
Human Rac1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human Rac1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7552-96
Supplier:  ANTIBODIES.COM