Order Entry
Netherlands
ContactUsLinkComponent
Human RhD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RhD in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8142-96
Supplier:  ANTIBODIES.COM
New Product
Human RhD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RhD in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8142-96
Supplier:  ANTIBODIES.COM