Order Entry
Netherlands
ContactUsLinkComponent
Mouse ATP1B3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse ATP1B3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6123-96
Supplier:  ANTIBODIES.COM
New Product
Mouse ATP1B3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse ATP1B3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6123-96
Supplier:  ANTIBODIES.COM