Order Entry
Netherlands
ContactUsLinkComponent
Mouse Chromogranin C/SGII ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Chromogranin C/SGII in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7281-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Chromogranin C/SGII ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Chromogranin C/SGII in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7281-96
Supplier:  ANTIBODIES.COM