Order Entry
Netherlands
ContactUsLinkComponent
Human 15 Lipoxygenase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human 15 Lipoxygenase 1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7978-96
Supplier:  ANTIBODIES.COM
New Product
Human 15 Lipoxygenase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human 15 Lipoxygenase 1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7978-96
Supplier:  ANTIBODIES.COM