Order Entry
Netherlands
ContactUsLinkComponent
Zebrafish IL-8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of zebrafish IL-8 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82431-96
Supplier:  ANTIBODIES.COM
New Product
Zebrafish IL-8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of zebrafish IL-8 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA82431-96
Supplier:  ANTIBODIES.COM