Order Entry
Netherlands
ContactUsLinkComponent
Mouse FNDC5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse FNDC5 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8107-96
Supplier:  ANTIBODIES.COM
New Product
Mouse FNDC5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse FNDC5 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8107-96
Supplier:  ANTIBODIES.COM