Order Entry
Netherlands
ContactUsLinkComponent
Mouse cAMP Protein Kinase Catalytic Subunit alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for th e in vitro quantitative determination of mouse cAMP Protein Kinase Catalytic Subunit alpha in serum, plasma, tissue h 1 * 96 Tests
Catalog # ANTIA75752-96
Supplier:  ANTIBODIES.COM
New Product
Mouse cAMP Protein Kinase Catalytic Subunit alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for th e in vitro quantitative determination of mouse cAMP Protein Kinase Catalytic Subunit alpha in serum, plasma, tissue h 1 * 96 Tests
Catalog # ANTIA75752-96
Supplier:  ANTIBODIES.COM