Order Entry
Netherlands
ContactUsLinkComponent
Mouse ITK/EMT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse ITK/EMT in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA7034-96
New Product
Mouse ITK/EMT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse ITK/EMT in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA7034-96