Order Entry
Netherlands
ContactUsLinkComponent
Mouse Vasorin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse Vasorin in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7812-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Vasorin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse Vasorin in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7812-96
Supplier:  ANTIBODIES.COM