Order Entry
Netherlands
Orders LinkContactUsLinkComponent
1-(2-(Methylamino)phenyl)ethanone 95%

1-(2-(Methylamino)phenyl)ethanone 95%

Bottle
Catalog # APOSOR929725-5G
New Product
Supplier:   Apollo Scientific
1-(2-(Methylamino)phenyl)ethanone 95%
Catalog # APOSOR929725-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  1859-75-2
1-(2-(Methylamino)phenyl)ethanone 95%

Bottle

CAS Number:  1859-75-2
Price
1 210,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:1859-75-2
  • Formula:C₉H₁₁NO
  • MDL Number:MFCD19159692
  • Molecular Weight:149.19 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications