Order Entry
Netherlands
Orders LinkContactUsLinkComponent
1-(2-(Methylamino)phenyl)ethanone 95%

1-(2-(Methylamino)phenyl)ethanone 95%

Bottle
Catalog # APOSOR929725-250MG
New Product
Supplier:   Apollo Scientific
1-(2-(Methylamino)phenyl)ethanone 95%
Catalog # APOSOR929725-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1859-75-2
1-(2-(Methylamino)phenyl)ethanone 95%

Bottle

CAS Number:  1859-75-2
Price
123,00 €
Log in for availability
1 * 250 mg
Product Details & Documents
  • CAS Number:1859-75-2
  • Formula:C₉H₁₁NO
  • MDL Number:MFCD19159692
  • Molecular Weight:149.19 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.