,,7 components: 1000mg/l each of Fe Mo K Mg Ca Na S in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM115428L1
Supplier: CUSTOM MADE CHEMICALS LAB